Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Registrieren
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Bottles
Glass Bottles
Reservoir bottles for automatic burettes
Reservoir bottles for automatic burettes
Reservoir bottles for automatic burettes
Catalog # HECH41917048
Supplier:
GLASWARENFABRIK KARL HECHT
Expand image
Reservoir bottles for automatic burettes
Catalog # HECH41917048
Supplier:
GLASWARENFABRIK KARL HECHT
Specifications
Capacity:
1000 ml
Colour:
Amber
Description:
Reservoir bottle with standard ground joint NS 29/32
Height:
200 mm
Pk:
1
Specifications
About this item
Produktdetails
Produktdetails