ContactUsLinkComponent
Allihn condensers with standard ground joints
Allihn condensers with standard ground joints
Catalog # HECH41987040
undefined
Allihn condensers with standard ground joints
Catalog # HECH41987040
Specifications
  • Cone size:
    29/32 NS
  • Jacket length:
    250 mm
  • Socket size:
    29/32 NS
  • Pk:
    1
Specifications
About this item
DURAN®, borosilicate glass 3.3.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
For condensing vapours. With standard ground joint cone and sleeve. Water connection via glass olive.
Certifications: Compliant with DIN 12581