Orders LinkContactUsLinkComponent
Analogue stopwatch
Analogue stopwatch
Stopwatch, 0 - 30 min
Catalog # HECH41978010
Analogue stopwatch
Catalog # HECH41978010
Analogue stopwatch
Analogue stopwatch
Stopwatch, 0 - 30 min
Price
393.00 SFr.
Log in for availability
1 * 1 ST
Product Details & Documents
Solid metal stopwatch with 7 jewels.
  • Graduation 0 to 60 in 1/5 seconds and 1/100 minutes
  • Timing capacity: Up to 30 minutes
  • Impact, dust and water resistant
Specifications
  • Description:
    Stopwatch, 0 - 30 min