PIG® spill response bag
: NEW PIG
PIG absorbents in the ideal spill kit bag - hi-viz, water-resistant, and easy to close, hang and carry.
- Durable, water-resistant bag with hook-and-loop closure keeps contents clean and dry
- PIG aborbent socks stop spreading spills; PIG mat absorbs quickly
- Temporary disposal bags help make cleanup easier
- Bright-yellow bag with bold Spill Kit lettering for quick identification
- Strap for easy carrying and hanging
Compact and easy to store kit is great for use in trucks or equipment.
Caution: To ensure effectiveness and your safety, we recommend that you conduct compatibility and absorption testing of your chemicals with PIG® HAZ-MAT products prior to purchase.
PIG absorbents in the ideal spill kit bag - hi-viz, water-resistant, and easy to close, hang and carry.
- Durable, water-resistant bag with hook-and-loop closure keeps contents clean and dry
- PIG aborbent socks stop spreading spills; PIG mat absorbs quickly
- Temporary disposal bags help make cleanup easier
- Bright-yellow bag with bold Spill Kit lettering for quick identification
- Strap for easy carrying and hanging
Compact and easy to store kit is great for use in trucks or equipment.
Caution: To ensure effectiveness and your safety, we recommend that you conduct compatibility and absorption testing of your chemicals with PIG® HAZ-MAT products prior to purchase.
Product Family Options
Universal
- DescriptionAbsorbencyW×D×H
- Spill kit, includes 4 absorbent socks, 28 mat pads and 5 disposal bags34,7 L150×430×520 mm1 * 1 STSFr. 158.00
HAZ-MAT
- DescriptionAbsorbencyW×D×H
- Spill kit, includes 4 absorbent socks, 28 mat pads and 5 disposal bags34,7 L150×430×520 mm1 * 1 KITSFr. 217.00
Oil-Only
- DescriptionAbsorbencyW×D×H
- Spill kit, includes 4 absorbent socks, 28 mat pads and 5 disposal bags38,5 L150×430×520 mm1 * 1 STSFr. 181.00