Orders LinkContactUsLinkComponent
Solid phase extraction cartridges, Bond Elut™ Focus
Solid phase extraction cartridges, Bond Elut™ Focus
Supplier:   VARIAN
Solid phase extraction cartridges, Bond Elut™ Focus
Supplier:  VARIAN
Certificates
Solid phase extraction cartridges, Bond Elut™ Focus
4 Options Available Below
Product Details & Documents
Bond Elut™ Focus solid phase extraction cartridges with enhanced polar recovery. Features a unique amide functionalised polar-enhanced sorbent technology that delivers outstanding retention for polar and non-polar analytes.
  • Simple, universal method reduces method validation times
  • 'Power Rinse' delivers cleaner extracts, improving sensitivity
  • Hydrogen bond donor capability increases retention of basic analytes
Rigorous washes using 10 to 20% ACN and MeOH can be used to aggressively remove endogenous matrix interferences. For very polar analytes that are held by strictly polar interactions.
Product Family Options
Product Information
  • Phase
    Pore size
    Particle Size
    Capacity
    Bed weight
    Availability
    Price
  • VARIA5306021
    Straight barrel cartridge
    Straight barrel cartridge
    Focus
    120 Å
    44 µm
    3 ml
    20 mg
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST
  • VARIA5606050
    Straight barrel cartridge
    Straight barrel cartridge
    Focus
    120 Å
    44 µm
    6 ml
    50 mg
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST
  • VARIA5106010
    Straight barrel cartridge
    Straight barrel cartridge
    Focus
    120 Å
    44 µm
    1 ml
    10 mg
    Log in for availability
    Log in for availability
    1 * 3 ST
    1 * 3 ST
  • VARIA5306022
    Straight barrel cartridge
    Straight barrel cartridge
    Focus
    120 Å
    44 µm
    3 ml
    60 mg
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST
Accessories
Product Information
  • Catalog #
    Filling
    Availability
    Price
  • 96-round well plate for SPE, Focus™
    96-round well plate for SPE, Focus™
    VARIA59660
    10 mg
    Log in for availability
    1 * 1 ST