Dishes, evaporating, flat or round Bottom, with spout
Fournisseur: Kavalier
Certificats
À propos de cet article
Made of borosilicate glass 3.3.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
For a wide range of applications. With flat bottom or rund bottom.
Frequently Bought Together