Orders LinkContactUsLinkComponent
Section lifters
Section lifters
Supplier:   RSG
Section lifters
Supplier:  RSG
Certificates
Section lifters
3 Options Available Below
Product Details & Documents
Section lifters with flexible blade and handle made of stainless steel.
Documents
Product Family Options
Product Information
  • Length
    Blade length
    Blade width
    Availability
    Price
  • 150 mm
    50 mm
    20 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - SFr. 74.10
    SFr. 74.10
    1 * 5 ST
       SFr. 74.10
  • 150 mm
    50 mm
    15 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - SFr. 73.70
    SFr. 73.70
    1 * 5 ST
       SFr. 73.70
  • 150 mm
    50 mm
    10 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - SFr. 72.90
    SFr. 72.90
    1 * 5 ST
       SFr. 72.90