DSS (Disuccinimidyl suberate)
Supplier: G-Biosciences
Certificates
About this item
CAS Number:68528-80-3
:
Formula:C₁₆H₂₀N₂O₈
MDL Number:MFCD00049059
:368.34 g/mol
Specification Test Results
- Spacer Arm (Å):11.4
- Reactive Toward:Primary Amines
- Membrane Permeable:Yes
- Water Soluble:No
- Cleavable/ Reversible:No
Specification Test Results
Frequently Bought Together