
5-Decyn-1-ol ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:68274-97-5
:
Formula:C₁₀H₁₈O
MDL Number:MFCD00464353
:154.25 g/mol
Boiling Pt:238 °C
Density:0.861
EINECS:269-529-4
:GHS07
Storage Temperature:Ambient
Frequently Bought Together