ContactUsLinkComponent
PIG® SpillBlocker® Dike, rough surface
PIG® SpillBlocker® Dike, rough surface
Catalog # NEWPPLR230
Supplier:  NEW PIG
undefined
PIG® SpillBlocker® Dike, rough surface
Catalog # NEWPPLR230
Supplier:  NEW PIG
Specifications
  • Description:
    Dike, rough surface
  • W×D×H:
    50×3000×60 mm
  • Packed:
    1 each
  • Pk:
    1
Specifications
About this item
Spills on asphalt can seep underneath standard spill dikes. Stop them in their tracks with our rough surface dike.
  • Exclusive New Pig formulation features a tear-resistant top layer with a super-tacky bottom layer that seals tight to rough surfaces and blocks liquids
  • Bottom layer "seeps" into surface cracks to form a liquid-tight seal
  • Fast protection for when a spill is heading right for your drain on a rough surface; just set dike down with sticky side away from surface, turn dike over and press into place
  • Confines and diverts fluids without absorbing them, making cleanup or reclamation simpler
  • 6cm height contains spills up to 5cm deep
  • Polyurethane material resists water, oils and many chemicals
  • High-visibility yellow top colour draws attention to spill for increased safety
Light-weight dike is easy to handle and deploy. Intended for one-time, limited term use; ideal for containing hazardous or other liquids on rough surfaces
Caution: Due to the nature of softer urethane, the physical properties of these products may change over time with exposure to certain environmental conditions like temperature, humidity and UV radiation. Typical life expectancy is 5 years. Please inspect the stored product regularly to ensure it is in a usable state. For more details and for chemical compatibility, please call Technical Services.