Pesticide reference standard, Piperonyl butoxide 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Katalognr. EHERL16240000CY
Anbieter: EHRENSTORFER
CAS-Nummer:
Technische Daten
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:51-03-6
- Description:Piperonyl butoxide 10 µg/ml in cyclohexane
- Environmentally Preferable:
- Formula:C₁₉H₃₀O₅
- Description:Piperonyl butoxide