Order Entry
Switzerland
ContactUsLinkComponent
Dodecenylsuccinic anhydride (mixture of isomers) hard for embedding, Supelco®
Irritant
Dodecenylsuccinic anhydride (mixture of isomers) hard for embedding, Supelco®
Catalog # 45346-250ML-F
CAS Number:  
undefined
Irritant
Dodecenylsuccinic anhydride (mixture of isomers) hard for embedding, Supelco®
Catalog # 45346-250ML-F
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 mL
  • Seq:
    2590

Specifications

About this item

 :
 :266.38 g/mol
 :M-Clarity™ quality level = MQ100
EINECS:246-917-1
 :GHS07
CAS Number:25377-73-5
 :26544-38-7
Formula:C₁₆H₂₆O₃
MDL Number:MFCD00036197