Order Entry
Switzerland
ContactUsLinkComponent
 
 
Dodecenylsuccinic anhydride (mixture of isomers), Sigma-Aldrich®
  8.22049.0250
 :  Merck
 :  
 
Dodecenylsuccinic anhydride (mixture of isomers), Sigma-Aldrich®
  8.22049.0250
 :  Merck
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    250 ml
  • Seq:
    2590

 

 

  • Solubility:
    (20 °C) insoluble
  • Molar Mass:
    266.38 g/mol
  • Boiling Point:
    200 °C (27 hPa)
  • Vapor Pressure:
    26 hPa (200 °C)
  • Flash Point:
    127 °C
  • Refractive Index:
    1.4745 (25 °C, 589 nm)
  • Density:
    1.01 g/cm3 (20 °C)
  • pH:
    2 - 4 ( 50 g/l, H2O, 20 °C) (slurry)

 

 

 :
 :266.38 g/mol
 :M-Clarity™ quality level = MQ200
 :246-917-1
 :GHS07
 :Ambient
 :25377-73-5
 :26544-38-7
 :C₁₆H₂₆O₃
 :MFCD00036197