Order Entry
Switzerland
Orders LinkContactUsLinkComponent
1,3-Dihydroxyacetone dimer ≥97%

1,3-Dihydroxyacetone dimer ≥97%

1,3-Dihydroxyacetone dimer ≥97%
Certificates
1,3-Dihydroxyacetone dimer ≥97%
1,3-Dihydroxyacetone dimer ≥97%

3 Options Available Below

Product Details & Documents

Purity: 97% CAS No.: 62147-49-3 Molecular Formula: C6H12O6 Formula Weight: 180.16 Storage Temperature: 2°C to 8°C Melting Point: ca 75-82° Beilstein Reference: 112910 Merck Reference: 14,3182 MDL No.: MFCD00051019

Synonyms:  2,5-Dihydroxy-1,4-dioxane-2,5-dimethanol
  • CAS Number:62147-49-3
  • Formula:C₆H₁₂O₆
  • MDL Number:MFCD00051019
  • Molecular Weight:180.16 g/mol
  • Merck Index:14,3182
  • Melting Pt:∼ 75…82 °C
  • EINECS:612-947-2
Product Family Options

Product Information