Order Entry
Switzerland
ContactUsLinkComponent
Pesticide reference standard, 2,3,5,6-Tetrachlorophenyl acetate 10 µg/ml in isooctane, Dr Ehrenstorfer
Pesticide reference standard, 2,3,5,6-Tetrachlorophenyl acetate 10 µg/ml in isooctane, Dr Ehrenstorfer
Catalog # EHERL17376200IO
Supplier:  EHRENSTORFER
CAS Number:  
Pesticide reference standard, 2,3,5,6-Tetrachlorophenyl acetate 10 µg/ml in isooctane, Dr Ehrenstorfer
Catalog # EHERL17376200IO
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    61925-90-4
  • Description:
    2,3,5,6-Tetrachlorophenyl acetate 10 µg/ml in isooctane
  • Formula:
    C₈H₄Cl₄O₂
  • Description:
    2,3,5,6-Tetrachlorophenyl acetate

Specifications

About this item