Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Progesterone

Glass bottle for solids, 25 g
Catalog # MOLEM71452029
Marketsource
Supplier:   Molekula
Progesterone
Catalog # MOLEM71452029
Supplier:  Molekula
Marketsource
CAS Number:  57-83-0
Progesterone

Glass bottle for solids, 25 g

CAS Number:  57-83-0
Price
Inquire For Price
Log in for availability
1 * 25 g
Product Details & Documents
  • CAS Number:57-83-0
  • Formula:C₂₁H₃₀O₂
  • MDL Number:MFCD00003658
  • EINECS:200-350-6
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.