Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Progesterone
Progesterone
Progesterone
Progesterone
Catalog # CHMCCBL70
Supplier:
Chemicon
CAS Number:
Bild vergrößern
Progesterone
Catalog # CHMCCBL70
Supplier:
Chemicon
CAS Number:
Specifications
Pack type:
Glass bottle for solids
Pk:
1 mg
Seq:
2070
Specifications
About this item
Produktdetails
:
EINECS:
200-350-6
CAS Number:
57-83-0
Formula:
C₂₁H₃₀O₂
MDL Number:
MFCD00003658
Produktdetails