Progesterone, Millipore®
Glass bottle for solids, 25 g
Catalog # 5341-25
Supplier: Merck Millipore (Calbiochem)
Progesterone, Millipore®
Catalog # 5341-25
Supplier: Merck Millipore (Calbiochem)
CAS Number: 57-83-0
Jump to:
Product Details & Documents
⁴-Pregnene-3,20-dione
Steroid hormone secreted by the corpus luteum during the latter half of the menstrual cycle. Binds to the progesterone receptor in the nucleus to influence transcription.
- CAS Number:57-83-0
- Formula:C₂₁H₃₀O₂
- MDL Number:MFCD00003658
- EINECS:200-350-6
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



