Order Entry
Switzerland
ContactUsLinkComponent
Organic reference standard, Ethylbenzene 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Organic reference standard, Ethylbenzene 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERL13320000CY
Supplier:  EHRENSTORFER
CAS Number:  
Organic reference standard, Ethylbenzene 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERL13320000CY
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    100-41-4
  • Description:
    Ethylbenzene 10 µg/ml in cyclohexane
  • Formula:
    C₈H₁₀
  • Description:
    Ethylbenzene

Specifications

About this item