Adenosine-5'-triphosphate disodium salt (ATP disodium salt) hydrate ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:34369-07-8
:
Formula:C₁₀H₁₄N₅Na₂O₁₃P₃·xH₂O
MDL Number:MFCD00080339
:551.14 g/mol (anhy)
Merck Index:14,00155
Storage Temperature:Refrigerator
EINECS:213-579-1
Specification Test Results
- Appearance:White to Almost white powder to crystaline
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 % (calcd.on anh.substance)
- abs-ratio (a/b):0.78 to 0.82 (E250/E260)
- abs-ratio (2a/2b):0.13 to 0.17 (E280/E260)
Specification Test Results
Frequently Bought Together