Order Entry
Switzerland
Orders LinkContactUsLinkComponent
DIPSO 99%
DIPSO 99%
Certificates
DIPSO 99%

2 Options Available Below

Product Details & Documents

Purity: 99% CAS No.: 68399-80-4 Molecular Formula: C7H17NO6S Formula Weight: 261.29 Beilstein Reference: 2105454 MDL No.: MFCD00038353

Synonyms:  3-(Bis(2-hydroxyethyl)amino)-2-hydroxypropane-1-sulphonic acid, DIPSO, 3-[Bis(2-hydroxyethyl)amino]-2-hydroxypropanesulphonic acid
  • CAS Number:68399-80-4
  • Formula:C₇H₁₇NO₆S
  • MDL Number:MFCD00038353
  • Molecular Weight:261.29 g/mol
  • Storage Temperature:Ambient
  • EINECS:269-992-2
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.