Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Enzymes
Ribonuclease A
Ribonuclease A
Ribonuclease A
Catalog # CHMCMAB3812
Supplier:
Chemicon
CAS Number:
Expand image
Ribonuclease A
Catalog # CHMCMAB3812
Supplier:
Chemicon
CAS Number:
Specifications
Pk:
100 µG
CAS No.:
9001-99-4
Storage conditions:
Freezer
MDL No.:
MFCD00132184
Enzyme name:
Ribonuclease A
Enzyme synonyms:
RNase A
Specifications
About this item
Product Details
Product Details