Order Entry
Switzerland
ContactUsLinkComponent
Tin(II) chloride dihydrate 98-103%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Corrosive
Irritant
Tin(II) chloride dihydrate 98-103%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Corrosive
Irritant
Tin(II) chloride dihydrate 98-103%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    98.0 - 103.0%
  • Assay (SnCl₂·2H₂O):
    98.0 - 103.0 %
  • Solubility in HCl:
    Passes Test
  • Sulfate (SO₄):
    Passes Test
  • Calcium (Ca):
    ≤0.005 %
  • Iron (Fe):
    ≤0.003 %
  • Lead (Pb):
    ≤0.01 %
  • Magnesium (Mg):
    ≤0.01 %
  • Potassium (K):
    ≤0.005 %
  • Sodium (Na):
    ≤0.010 %
  • Trace Impurities - Arsenic (As):
    ≤2.000 ppm
  • Trace Impurities - Mercury (Hg):
    ≤0.050 ppm

Specification Test Results

Related Information

Frequently Bought Together