Order Entry
Switzerland
ContactUsLinkComponent
Molecular sieve 13X (1.0 nm, 10 Å)
Molecular sieve 13X (1.0 nm, 10 Å)
Catalog # SUPL20307
Supplier:  Merck
CAS Number:  
Molecular sieve 13X (1.0 nm, 10 Å)
Catalog # SUPL20307
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g
  • Seq:
    3368

Specifications

About this item

 :
CAS Number:63231-69-6
 :1318-02-1
MDL Number:MFCD00164506