Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Molecular sieve 13X (1.0 nm, 10 Å)
Molecular sieve 13X (1.0 nm, 10 Å)
Molecular sieve 13X (1.0 nm, 10 Å)
Molecular sieve 13X (1.0 nm, 10 Å)
Catalog # SUPL20305
Supplier:
Merck
CAS Number:
Expand image
Molecular sieve 13X (1.0 nm, 10 Å)
Catalog # SUPL20305
Supplier:
Merck
CAS Number:
Specifications
Pk:
50 g
Seq:
3368
Specifications
About this item
Product Details
:
CAS Number:
63231-69-6
:
1318-02-1
MDL Number:
MFCD00164506
Product Details