Order Entry
Switzerland
ContactUsLinkComponent
Potassium hydrogen phthalate 99.95-100.05% (dry basis), crystals, BAKER ANALYZED® ACS, J.T.Baker®
Potassium hydrogen phthalate 99.95-100.05% (dry basis), crystals, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Potassium hydrogen phthalate 99.95-100.05% (dry basis), crystals, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Meets ACS Reagent Chemical Requirements:
    99.95 - 100.05%
  • For Laboratory,Research,or Manufacturing Use:
    ≤0.005%
  • Assay (1-KOCOC₆H₄-2-COOH) (dried basis):
    99.95 - 100.05 %
  • Insoluble Matter:
    ≤0.005 %
  • pH of 0.05M Solution at 25°C:
    4.00 - 4.02
  • Chlorinated Compounds (as Cl):
    ≤0.003 %
  • Sulfur Compounds (as S):
    ≤0.002 %
  • Sodium (Na):
    ≤0.005 %
  • Trace Impurities - ACS - Heavy Metals (as Pb):
    ≤5 ppm
  • Trace Impurities - Iron (Fe):
    ≤5 ppm

Specification Test Results

Related Information

Frequently Bought Together