About this item
Ideal for the use in production and laboratory, this budget-priced, basic model has a generously proportioned seat and a high backrest. With black synthetic leather.
- Washable, impervious to oil and disinfecting agents
Unitec Basic
Large, ergonomically designed backrest to take the strain off the spine and muscles with integrated lumbar support
Backrest height adjustment convenient and easy to handle in sitting position
Healthy posture due to a hump providing support for the back in the lumbar region
Ergonomically designed seat with rounded front edge, which prevents blocked circulation in the thighs
Stepless seat height setting supported by safety gas spring with protective cover
Permanent contact backrest: this mechanism ensures that the backrest responds immediately to any movements of the upper body by following them
Environment-friendly: the materials used are recyclable
5 black star base made from plastic for high stability, longetivity and safety; the flat design prevents stumbling hasard
Unitec ESD
The metal components have a conductive coating
The high-comfort upholstery is also conducive
With conductive glides or castors
Seat and backrest made from ESD fabric upholstery: resistant, breathable and comfortable
5 star base made from steel shape tube, conductive, powder-coated for high stability, longetivity and safety; the black flat design prevents stumbling hasard
Large, ergonomically designed backrest to take the strain off the spine and muscles with integrated lumbar support
Backrest height adjustment convenient and easy to handle in sitting position
Healthy posture due to a hump providing support for the back in the lumbar region
Ergonomically designed seat with rounded front edge, which prevents blocked circulation in the thighs
Stepless seat height setting supported by safety gas spring with protective cover
Permanent contact backrest: this mechanism ensures that the backrest responds immediately to any movements of the upper body by following them
Environment-friendly: the materials used are recyclable
Confirms with quality management system DIN EN ISO 9001 standards
Complies with regulation EWG Nr. 1836/93 and environmental management system DIN ISO 14001 standards
Unitec 1,2,3 ESD: exceeds the requirements of ESD protection according to standard DIN EN 61340-5-1, 100 % ESD protection
Specifications
- Catalog No:
- BIMO9651-3000
- BIMO9651-2000
- BIMO9653E-9801
- BIMO9653-0562
- BIMO9651-CI01
- 132-1083
- BIMO9653-3000
- BIMO9650-2000
- BIMO9653-CI01
- BIMO9650-CI01
- 132-1082
- BIMO9650-0551
- BIMO9651E-9801
- 132-1080
- 132-1078
- BIMO9653E-9802
- BIMO9650E-9801
- BIMO9653-2000
- BIMO9650E-2571
- BIMO9650-3000
- Seat dimensions:
- -
- -
- -
- -
- -
- -
- -
- -
- -
- -
- -