Orders LinkContactUsLinkComponent
Deactivated/silinised vials
Deactivated/silinised vials
Deactivated/silinised vials
Certificates
Deactivated/silinised vials
Deactivated/silinised vials
4 Options Available Below
Product Details & Documents
These vials made of PP are used for pH sensitive samples, sodium or heavy metals analysis.
  • Translucent
  • Good chemical resistance
  • Can be used with crimp or snap caps
Vials with glass insert are used for economical and effective microsampling. The PP body has a glass flanged insert molded to the inside, so the sample comes in contact only with the 100 µl glass insert and the septum.
Product Family Options
Product Information
  • Capacity
    Ø×H
    Cap included
    Cap type
    Availability
    Price
  • HEWL9301-0977
    Vials, with glass insert
    Vials, with glass insert
    0,25 ml
    12×32 mm
    No
    Crimp/snap
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST
  • HEWL5188-2788
    Vials, certified
    Vials, certified
    0,25 ml
    12×32 mm
    No
    Crimp/snap
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST
  • HEWL5188-5390
    Vials, with glass insert
    Vials, with glass insert
    0,25 ml
    12×32 mm
    No
    Screw
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST
  • Vials
    1,00 ml
    12×32 mm
    No
    Crimp/snap
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST
This product has been blocked by your organization. Please contact your purchasing department for more information