Orders LinkContactUsLinkComponent
Bellow connectors
Bellow connectors
Supplier:   COWIE
Bellow connectors
Supplier:  COWIE
Certificates
Bellow connectors
Bellow connectors
5 Options Available Below
Product Details & Documents
Bellow connector, PTFE, for use as a flexible or elastic connector to prevent pressure and stresses, and to dampen vibrations in standard ground joints.
  • Inert
  • Temperature resistant up to 280 °C
  • Excellent chemical resistance
With ground joints on cone and socket. Sturdy construction with trapezoidal sealing rings, which give medium to high vacuum performance.
Product Family Options
Product Information
  • Cone size
    Total length
    Socket size
    Availability
    Price
  • 14/35 NS
    115 mm
    14/35 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 81.70
    SFr. 81.70
    1 * 1 ST
       SFr. 81.70
  • 19/38 NS
    120 mm
    19/38 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 82.60
    SFr. 82.60
    1 * 1 ST
       SFr. 82.60
  • 24/40 NS
    130 mm
    24/40 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 114.00
    SFr. 114.00
    1 * 1 ST
       SFr. 114.00
  • 29/42 NS
    140 mm
    29/42 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 127.00
    SFr. 127.00
    1 * 1 ST
       SFr. 127.00
  • 19/22 NS
    90 mm
    19/22 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 97.90
    SFr. 97.90
    1 * 1 ST
       SFr. 97.90