Bottle pourers
Supplier: COWIE
Jump to:
About this item
Manufactured in pure, FDA approved, PTFE.
- Totally chemically resistant and inert
- To fit GL45, 38, 32 and 30 screw neck bottles
- State cap colour required, blue, red or green
Permits direct pouring in a safe and consistent manner.
4 Options Available Below
Product Details & Documents
Manufactured in pure, FDA approved, PTFE.
- Totally chemically resistant and inert
- To fit GL45, 38, 32 and 30 screw neck bottles
- State cap colour required, blue, red or green
Permits direct pouring in a safe and consistent manner.
Product Family Options
Product Information
- TypeAvailabilityPrice
- COWI028.030Bottle pourer, PTFE, blue
- COWI028.032Bottle pourer, PTFE, blue
- COWI028.038Bottle pourer, PTFE, blue
- COWI028.045BLUEBottle pourer, PTFE, blue
This product has been blocked by your organization. Please contact your purchasing department for more information