Powder funnels
Anbieter: GLASWARENFABRIK KARL HECHT
Zertifikate
Über diesen Artikel
Powder funnels, made of PP, translucent. For decanting powdery materials. Also suitable for liquids. Cone angle 60°.
- Excellent chemical resistance
- Excellent temperature resistance
- Autoclavable
Frequently Bought Together