Bottles, wide neck, with screw cap
Catalog # HECH49101042
Supplier: GLASWARENFABRIK KARL HECHT
Specifications
- Thread:GL 65
- Capacity:2000 ml
- Pk:15
Specifications
About this item
PE, translucent.
- Good chemical resistance
- Good temperature resistance
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904