Orders LinkContactUsLinkComponent
Test tubes with rim
Test tubes with rim
Test tubes with rim
Certificates
Test tubes with rim
4 Options Available Below
Product Details & Documents
PP, translucent or PS, clear, with rim.
  • PS: Good thermal and chemical resistance
  • PP: Excellent thermal and chemical resistance, autoclavable
  • Robust, economical
Universal tubes for a wide range of applications.
Product Family Options
Product Information
  • Capacity
    Length
    Ext.Ø
    Availability
    Price
  • 5 ml
    86 mm
    12 mm
    Log in for availability
    Log in for availability
    1 * 4.000 ST
    1 * 4.000 ST
  • 10 ml
    100 mm
    16 mm
    Log in for availability
    Log in for availability
    1 * 2.000 ST
    1 * 2.000 ST
  • 5 ml
    86 mm
    12 mm
    Log in for availability
    Log in for availability
    1 * 4.000 ST
    1 * 4.000 ST - SFr. 353.00
    SFr. 353.00
    1 * 4.000 ST
       SFr. 353.00
  • 10 ml
    100 mm
    16 mm
    Log in for availability
    Log in for availability
    1 * 2.000 ST
    1 * 2.000 ST - SFr. 258.00
    SFr. 258.00
    1 * 2.000 ST
       SFr. 258.00