Safety funnels
Supplier: DUPERTHAL
Certificates
About this item
Stainless steel safety funnels with hinged lid and butress thread S 56×4.
- Overfilling protection
- Lid reduces the escaping vapours
- Flame arrester made from stainless steel (simple to interchange)
Frequently Bought Together