Order Entry
Switzerland
ContactUsLinkComponent
(R)-(-)-2,5-Dihydro-3,6-dimethoxy-2-isopropylpyrazine for synthesis, Sigma-Aldrich®
(R)-(-)-2,5-Dihydro-3,6-dimethoxy-2-isopropylpyrazine for synthesis, Sigma-Aldrich®
Catalog # 8.18315.0100
Supplier:  Merck
CAS Number:  
(R)-(-)-2,5-Dihydro-3,6-dimethoxy-2-isopropylpyrazine for synthesis, Sigma-Aldrich®
Catalog # 8.18315.0100
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Solubility:
    (20 °C) insoluble
  • Molar Mass:
    184.23 g/mol
  • Boiling Point:
    100 - 102 °C (20 hPa)
  • Flash Point:
    85 °C
  • Density:
    1.03 g/cm3 (20 °C)

Specification Test Results

About this item

 :
 :184.24 g/mol
 :M-Clarity™ quality level = MQ200
CAS Number:109838-85-9
Formula:C₉H₁₆N₂O₂
MDL Number:MFCD00040565