4-(2-Pyridylazo)resorcinol 96%, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
CAS Number:1141-59-9
:
Formula:C₁₁H₉N₃O₂
MDL Number:MFCD00044237
:215.21 g/mol
Melting Pt:192...202 °C (lit.)
:M-Clarity™ quality level = MQ200
Storage Temperature:Room Temperature
EINECS:214-528-6
Specification Test Results
- Appearance (colour):Conforms to requirements
- Appearance (form):Powder
- Purity (HPLC area %):≥95.5 %
- Carbon content:58.6 - 64.2 %
- Nitrogen content:18.6 - 20.4 %
- Infrared spectrum:Conforms to structure
Specification Test Results
Frequently Bought Together