4-(2-Pyridylazo)resorcinol for synthesis, Sigma-Aldrich®
8.41016.0005
: Merck
:
- Pack type:Plastic bottle
- Pk:5 g
- Seq:3173
- Melting Point:187 - 189 °C
- Molar Mass:215.21 g/mol
- Synonyms:4-(2-Pyridinylazo)-1,3-dihydroxybenzene
- Hill Formula:C11H9N3O2
:
:215.21 g/mol
:M-Clarity™ quality level = MQ200
:214-528-6
:1141-59-9
:C₁₁H₉N₃O₂
:MFCD00044237