
[2-(2-Methoxyethoxy)ethoxy]acetic acid(3,6,9-trioxadecanoic acid) for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:16024-58-1
:
Formula:C₇H₁₄O₅
MDL Number:MFCD00044098
:178.1831 g/mol
ADR:8,III
:M-Clarity™ quality level = MQ200
EINECS:240-162-1
UN:3265
:GHS05
Storage Temperature:Ambient
Specification Test Results
- Molar Mass:178.18 g/mol
- Boiling Point:140 °C (2 mbar)
- Flash Point:160 °C
- Density:1.157 g/cm3
- pH:1.8 (100 g/l, H2O)
Specification Test Results
Frequently Bought Together