
1,4,7-Trimethyl-1,4,7-triazacyclononane ≥98.0% (by GC, titration analysis) stabilised
Supplier: TCI
Certificates
About this item
CAS Number:96556-05-7
:
Formula:C₉H₂₁N₃
MDL Number:MFCD00012359
:171.28 g/mol
Flash Pt:68°C
UN:3267
ADR:8,II
:Stabilised with sodium hydrogen carbonate (NaHCO₃)
:GHS05
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Colorless to Light orange to Yellow clear liquid
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together