Order Entry
Switzerland
ContactUsLinkComponent
Cyclohexane ≥99.8%, SupraSolv® for GC-MS
Flammable
Health
Irritant
Environment
Cyclohexane ≥99.8%, SupraSolv® for GC-MS
Supplier:  Merck
Certificates
undefined
Flammable
Health
Irritant
Environment
Cyclohexane ≥99.8%, SupraSolv® for GC-MS
Supplier:  Merck
Limited Time Sale
Select items below are currently on sale

Specification Test Results

  • Purity (GC):
    =99.8%
  • Identity(IR):
    Conforms
  • Evaporation Residue:
    = 3.0 mg/l
  • Water:
    = 0.01%
  • Color:
    =10 Hazen
  • GC/MSD (retention range n-undecane to n-tetracontane, scanning area 30-600 amu, individual signals (n-tetradecane standard)):
    = 3ng/ml
  • Suitability:
    Residue Analysis

Specification Test Results

Related Information

Frequently Bought Together