Double tube end fittings, BOLA
Anbieter: Bohlender
Zertifikate
Über diesen Artikel
PTFE, white.
- Temperature resistance: from -200 to +250 °C
- Universal chemical resistance
- FDA conform
With two male threads UNF ¼" 28 G.
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Anbieter: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Anbieter: VWR Chemicals
VWR®, Screw Neck Vials ND13
Anbieter: VWR Collection
VWR®, Screw Caps for Short Thread Vials ND9
Anbieter: VWR Collection
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Anbieter: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Anbieter: VWR Chemicals
VWR®, Snap Cap Vials
Anbieter: VWR Collection
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Anbieter: VWR Chemicals
VWR®, Short Thread Vials ND9
Anbieter: VWR Collection
VWR®, Crimp Neck Vials ND11
Anbieter: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Anbieter: VWR Chemicals
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Anbieter: VWR Chemicals
TLC plates, aluminium oxide, Alox
Anbieter: MACHEREY-NAGEL
Hand brushes and brooms, PP
Anbieter: PFENNIG REINIGUNGSTECHNIK
VWR®, Jars, Wide Mouth, with Screw Cap
Anbieter: VWR Collection
Miniature screw-in adaptors, BOLA
Anbieter: Bohlender