ContactUsLinkComponent
Beakers, low form, with spout
Beakers, low form, with spout
Catalog # WITG8.600.150
Supplier:  witeg Labortechnik
Supplier #: 8.600.150
undefined
Beakers, low form, with spout
Catalog # WITG8.600.150
Supplier:  witeg Labortechnik
Supplier #: 8.600.150
Log in for availability
Select an Option
55 mm
Select an Option
150 ml
Select an Option
80 mm
Select an Option
1
About this item
Quartz glass.
  • High thermal shock resistance with low thermal expansion
  • High transmission from the UV to the IR spectrum range
  • High chemical resistance and purity
  • High electrical insulation properties
  • High radiation resistance
  • High temperature resistance and softening temperature
The combination of the special thermal, electrical, chemical, mechanical and optical properties in a single material makes quartz glass indispensible for high-technology applications.
Frequently Bought Together