Bottles, wide neck, amber, with screw caps
Catalog # WITG5.895.002
Anbieter: witeg Labortechnik
Technische Daten
- Thread:GL 80
- Capacity:2500 ml
- Seq:02
- Pk:1
Technische Daten
Über diesen Artikel
Amber bottles, wide mouth with DIN.
- Good chemical and physical properties
- For applications that do not involve high thermal stresses
- Round thread in accordance with DIN 168
Ideal for sampling, storing and transporting powders and pastes. Amber coloured to protect light-sensitive materials.