Order Entry
Switzerland
ContactUsLinkComponent
 
Vacuum desiccators, complete
  WITG5.661.250
undefined
Vacuum desiccators, complete
  WITG5.661.250

 

  • Nominal size:
    250
  • Type:
    with adapters, side tube ST 24/29 and 2 PTFE-needle valve stopcocks, 250 mm Ø
  • Pk:
    1
  • Usable height:
    120 mm
  • Ø ext.×H:
    320×344 mm

 

 

Borosilicate glass 3.3. This vacuum desiccator has 10.5 litre capacity.

 : Supplied with two manometers, lid and side tube ST 24/29, PTFE needle valve stopcocks and with ring nut which has a safety coating.