ContactUsLinkComponent
PCR tubes, 0,2 ml, with flat cap
Catalog # BITX3423.AS.X
Supplier:  BIOTIX
undefined
PCR tubes, 0,2 ml, with flat cap
Catalog # BITX3423.AS.X
Supplier:  BIOTIX
Specifications
  • Packed:
    10 bags of 100/pack
  • Capacity:
    0,2 ml
  • Colour:
    Assorted
  • Description:
    with flat cap, sterile
  • Pk:
    1.000
Specifications
About this item
Made of high quality virgin polypropylene.
  • Excellent chemical resistance
  • Excellent temperature resistance
  • Certified free of DNA, DNase, RNase, and endotoxins (pyrogens)
PCR tubes feature uniform microthin walls for efficient heat transfer. Tubes and caps are compatible with most leading thermal cyclers. Caps fit perfectly and create a uniform tight seal that prevents sample evaporation in the thermal cycler.
Manufactured under stringent controls in an ISO 9001 certified facility. Every lot is tested for precision assuring maximum quality and compatibility. All raw materials are traceable.
Certifications: CE certified. IVD labeled.