ContactUsLinkComponent
Anion exchange HPLC columns, PRP-X100
Anion exchange HPLC columns, PRP-X100
Supplier:  HAMILTON BONADUZ
Certificates
undefined
Anion exchange HPLC columns, PRP-X100
Supplier:  HAMILTON BONADUZ
Specifications
  • Catalog No:
  • 554-2272
  • 554-2273
  • 554-2275
  • 554-2274
  • Max. pressure:
  • 350 bar
  • 350 bar
  • 350 bar
  • 350 bar
  • pH range:
  • 1 - 13
  • 1 - 13
  • 1 - 13
  • 1 - 13
Specifications
Guard cartridge holder
Product Information