HPLC columns, Acclaim™ Surfactant Plus
Supplier: Thermo Fisher Scientific
Certificates
About this item
These columns have been designed specifically for the HPLC separation of surfactants including anionic, non ionic, cationic and amphoteric used in a wide range of samples, such as customer products, pharmaceutical, food and beverages, environmental samples etc.
- Unique polarity provides significantly improved resolution compared with conventional C18 columns
- Designed for use with charged aerosol detectors and mass spectrometers
- Can be used with evaporative light scattering detectors, suppressed conductivity detectors and UV/Visible detectors
Particle type: Ultra-pure, spherical silica
Bonded phase: Mixed-mode reversed phase and anion exchange
Hydrophobicity: Moderate to low
Carbon load: 5%
pH range: 2,5 to 7,5
Frequently Bought Together