L(-)-Carnitine
Supplier: Biomol Enzo
Jump to:
About this item
Synonyms: Levocarnitine, L-Carnitine, Vitamin BT, (R)-3-Hydroxy-4-(trimethylammonio)butanoate, R)-(-)-3-Hydroxy-4-(trimethylammonio)butyrate
- CAS Number:541-15-1
- Formula:C₇H₁₅NO₃
- MDL Number:MFCD00038747
- Molecular Weight:161.20 g/mol
- EINECS:208-768-0
2 Options Available Below
Product Details & Documents
Synonyms: Levocarnitine, L-Carnitine, Vitamin BT, (R)-3-Hydroxy-4-(trimethylammonio)butanoate, R)-(-)-3-Hydroxy-4-(trimethylammonio)butyrate
- CAS Number:541-15-1
- Formula:C₇H₁₅NO₃
- MDL Number:MFCD00038747
- Molecular Weight:161.20 g/mol
- EINECS:208-768-0
Product Family Options
Product Information
- Pack typePkAvailabilityPrice



