Order Entry
Switzerland
ContactUsLinkComponent
Organic reference standard, 1-Methylfluoranthene 10 µg/ml in acetonitrile, Dr Ehrenstorfer
Organic reference standard, 1-Methylfluoranthene 10 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERL20892500AL
Supplier:  EHRENSTORFER
CAS Number:  
Organic reference standard, 1-Methylfluoranthene 10 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERL20892500AL
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    25889-60-5
  • Description:
    1-Methylfluoranthene 10 µg/ml in acetonitrile
  • Environmentally Preferable:
  • Formula:
    C₁₇H₁₂
  • Description:
    1-Methylfluoranthene

Specifications

About this item