Organic reference standard, (Aminomethyl)phosphonic acid 100 µg/ml in aqueous solution, Dr Ehrenstorfer
Catalog # EHERXA10205000WA
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:1066-51-9
- Description:(Aminomethyl)phosphonic acid 100 µg/ml in aqueous solution
- Formula:CH₆NOP
- Description:(Aminomethyl)phosphonic acid